Learn More
Ace Glass, Inc. Flask, recovery, heavy wall, 2L, 29/32
SDP
Catalog No. NC0557744
Encompass_Preferred
2L round bottom heavy wall recovery flask, with a single neck with a 29/32 joint, and a sloping side wall.
Catalog No. NC0557744 Supplier Ace Glass, Inc. Supplier No. 6892243
2L round bottom heavy wall recovery flask, with a single neck with a 29/32 joint, and a sloping side wall.