Learn More
CELLTREAT Scientific Products Flask Cap, Plug Seal (fits 75cm2 & 250mL), Sterile
SDP
Catalog No. 50202091
Encompass_Preferred
CELLTREAT Scientific Products Replacement Flask Cap. Replacement caps available for Tissue Culture Flasks. Plug seal cap fits 75cm¿ & 250mL flasks. Rnase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 5/bag.
Catalog No. 50-202-091 Supplier CELLTREAT Scientific Products Supplier No. 229396
CELLTREAT Scientific Products Replacement Flask Cap. Replacement caps available for Tissue Culture Flasks. Plug seal cap fits 75cm¿ & 250mL flasks. Rnase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 5/bag.