Learn More
CELLTREAT Scientific Products Flask Cap, Plug Seal (fits 25cm2 & 50mL), Sterile
SDP
Catalog No. 501117986
Encompass_Preferred
CELLTREAT Scientific Products Replacement Flask Cap. Replacement caps available for Tissue Culture Flasks. Plug seal cap fits 25cm¿ & 50mL flasks. Rnase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 5/bag.
Catalog No. 50-111-7986 Supplier CELLTREAT Scientific Products Supplier No. 229389
CELLTREAT Scientific Products Replacement Flask Cap. Replacement caps available for Tissue Culture Flasks. Plug seal cap fits 25cm¿ & 50mL flasks. Rnase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 5/bag.