Learn More
CELLTREAT Scientific Products Flask Cap, Vent (fits 12.5cm2 & 25mL), Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229393
Encompass_Preferred
CELLTREAT Scientific Products Replacement Flask Cap. Replacement caps available for Tissue Culture Flasks. Vent cap fits 12.5cm¿ & 25mL flasks. Rnase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 5/bag.
Catalog No. 50-202-088