Learn More
Chemglass Life Sciences 50mL 2-Neck Pear Shaped Flask, 1-CN 14/20 Outer, 1-SN 14/20 Outer Angled 20°
SDP
Catalog No. CG155814
Encompass_Preferred
50mL 2-Neck Pear Shaped Flask, 1-CN 14/20 Outer, 1-SN 14/20 Outer Angled 20¿. Pear shape, HEAVY WALL 2-neck flask with standard taper outer joints. Flasks with a 10/18 outer joint on the side neck are used with either jointed thermometers or CG-1079 inlet adapters.
Catalog No. CG155814 Supplier Chemglass Life Sciences Supplier No. CG155814
50mL 2-Neck Pear Shaped Flask, 1-CN 14/20 Outer, 1-SN 14/20 Outer Angled 20¿. Pear shape, HEAVY WALL 2-neck flask with standard taper outer joints. Flasks with a 10/18 outer joint on the side neck are used with either jointed thermometers or CG-1079 inlet adapters.