Learn More
Chemglass Life Sciences 500mL 3-Neck Round Bottom Flask (RBF) , 1-CN 24/40 Outer, 2-SN 24/40 Outer, 4mm Stpk Drain
SDP
Catalog No. CG152301
Encompass_Preferred
500mL 3-Neck Round Bottomed Flask, 1-CN 24/40 Outer, 2-SN 24/40 Outer, 4mm Stopcock Drain. Round bottom, HEAVY WALL, three neck flask with standard taper outer joints on the center and side necks. Lower outlet is a CG-424-05, 4mm straight bore PTFE stopcock.
Catalog No. CG152301 Supplier Chemglass Life Sciences Supplier No. CG152301
500mL 3-Neck Round Bottomed Flask, 1-CN 24/40 Outer, 2-SN 24/40 Outer, 4mm Stopcock Drain. Round bottom, HEAVY WALL, three neck flask with standard taper outer joints on the center and side necks. Lower outlet is a CG-424-05, 4mm straight bore PTFE stopcock.