Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, three neck, 200mL, 24/40 center, 24/40 sides
SDP
Supplier:  Ace Glass, Inc. 6944205
Encompass_Preferred
200mL heavy wall, round bottom flask with three vertical necks. Center neck and side necks have 24/40 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.
Catalog No. 50-973-680