Learn More
Ace Glass, Inc. Flask, round, heavy wall, three neck, 250mL, 14/20 center, 14/20 sides
SDP
Catalog No. 50976863
Encompass_Preferred
250mL round bottom, heavy wall flask with three necks for mechanical shock resistance. Flask features a 14/20 center neck and two 14/20 angled side necks.
Catalog No. 50-976-863 Supplier Ace Glass, Inc. Supplier No. 9465213
250mL round bottom, heavy wall flask with three necks for mechanical shock resistance. Flask features a 14/20 center neck and two 14/20 angled side necks.