Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, 250mL, 24/40 center, 24/40side
SDP
Catalog No. 50973593
Encompass_Preferred
250mL heavy wall round bottom flask with one center and one vertical side neck, both with 24/40 standard taper joints.
Catalog No. 50-973-593 Supplier Ace Glass, Inc. Supplier No. 6928208
250mL heavy wall round bottom flask with one center and one vertical side neck, both with 24/40 standard taper joints.