Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, 250mL, 29/42 center, 24/40side
SDP
Catalog No. 50973594
Encompass_Preferred
250mL heavy wall round bottom flask with one center neck with a 29/42 standard taper joint and one vertical side neck with a 24/40 standard taper joint.
Catalog No. 50-973-594 Supplier Ace Glass, Inc. Supplier No. 6928211
250mL heavy wall round bottom flask with one center neck with a 29/42 standard taper joint and one vertical side neck with a 24/40 standard taper joint.