Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, two neck, 500mL, 24/40 center, 10/30 side
SDP
Catalog No. 50978934
Encompass_Preferred
500mL two neck, round bottom heavy wall flask with standard joints. Center neck has a 24/40 joint, and the angled side neck has a 10/30 joint.
Catalog No. 50-978-934 Supplier Ace Glass, Inc. Supplier No. 6927220
500mL two neck, round bottom heavy wall flask with standard joints. Center neck has a 24/40 joint, and the angled side neck has a 10/30 joint.