Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, four neck, 500mL, 29/40 center, (2) 24/40, (1) #7 sides
SDP
Catalog No. 50973874
Encompass_Preferred
500mL heavy wall, round bottom flask with four necks, arranged three in-line, and one #7 Ace-Thred neck at 90. Center neck has 29/40 center joint, and angled side necks have 24/40 joints. Supplied with bushing and FETFE o-ring.
Catalog No. 50-973-874 Supplier Ace Glass, Inc. Supplier No. 6954276
500mL heavy wall, round bottom flask with four necks, arranged three in-line, and one #7 Ace-Thred neck at 90. Center neck has 29/40 center joint, and angled side necks have 24/40 joints. Supplied with bushing and FETFE o-ring.