Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, two neck, 500mL, 29/42 center, 24/40 side
SDP
Catalog No. 50973585
Encompass_Preferred
500mL two neck, round bottom heavy wall flask with standard joints. Center neck has a 29/42 joint, and the angled side neck has a 24/40 joint.
Catalog No. 50-973-585 Supplier Ace Glass, Inc. Supplier No. 6927229
500mL two neck, round bottom heavy wall flask with standard joints. Center neck has a 29/42 joint, and the angled side neck has a 24/40 joint.