Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, four neck, 500mL, 29/42 center, 29/42 sides
SDP
Catalog No. 50973833
Encompass_Preferred
500ml round bottom, heavy wall flask with four necks, arranged three in-line and one at 90. Center neck has a 29/42 joint, and angled side necks have 29/42 joints.
Catalog No. 50-973-833 Supplier Ace Glass, Inc. Supplier No. 6952206
500ml round bottom, heavy wall flask with four necks, arranged three in-line and one at 90. Center neck has a 29/42 joint, and angled side necks have 29/42 joints.