Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, three neck, 500mL, 45/50 center, 29/42 sides
SDP
Catalog No. 50973687
Encompass_Preferred
500mL heavy wall, round bottom flask with three vertical necks. Center neck has 45/50 joint, and side necks have 29/42 joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.
Catalog No. 50-973-687 Supplier Ace Glass, Inc. Supplier No. 6944229
500mL heavy wall, round bottom flask with three vertical necks. Center neck has 45/50 joint, and side necks have 29/42 joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.