Learn More
Ace Glass, Inc. Flask, round bottom, rotary evaporator, graduated, 250mL, 35/20
SDP
Catalog No. 50973542
Encompass_Preferred
250mL graduated, round bottom receiving flask for use with all rotary evaporators. Features a 35/20 spherical outer joint which can also mate to 35/25 ball joints. Graduated in 10mL increments.
Catalog No. 50-973-542 Supplier Ace Glass, Inc. Supplier No. 689305
250mL graduated, round bottom receiving flask for use with all rotary evaporators. Features a 35/20 spherical outer joint which can also mate to 35/25 ball joints. Graduated in 10mL increments.