Learn More
Ace Glass, Inc. Flask, round, 100mL, 14/20 center, 14/20 side
SDP
Catalog No. 50976850
Encompass_Preferred
100mL standard wall round bottom flask with (1) 14/20 standard taper center neck and (1) 14/20 standard taper angled side neck. Standard wall allows for more rapid heating.
Catalog No. 50-976-850 Supplier Ace Glass, Inc. Supplier No. 946410
100mL standard wall round bottom flask with (1) 14/20 standard taper center neck and (1) 14/20 standard taper angled side neck. Standard wall allows for more rapid heating.