Learn More
Ace Glass, Inc. Flask, round bottom, 100mL, 19/38 center, beaded tube port
SDP
Catalog No. 50978943
Encompass_Preferred
100mL round bottom, single neck flask with either a beaded tube end port or a beaded tube end well.
Catalog No. 50-978-943 Supplier Ace Glass, Inc. Supplier No. 693504
100mL round bottom, single neck flask with either a beaded tube end port or a beaded tube end well.