Learn More
Ace Glass, Inc. Flask, round, heavy duty, 100mL, 24/40 center, 14/20 side
SDP
Catalog No. 50976846
Encompass_Preferred
100mL heavy wall round bottom flask with a 24/40 center neck, and a 14/20 angled side neck. Heavy wall is approximately 30% heavier than regular wall flasks.
Catalog No. 50-976-846 Supplier Ace Glass, Inc. Supplier No. 9463211
100mL heavy wall round bottom flask with a 24/40 center neck, and a 14/20 angled side neck. Heavy wall is approximately 30% heavier than regular wall flasks.