Learn More
Ace Glass, Inc. Flask, round, 100mL, 24/40 center, 14/20 side
SDP
Catalog No. 50976842
Encompass_Preferred
100mL round bottom flask with a 24/40 center neck, and a 14/20 angled side neck.
Catalog No. 50-976-842 Supplier Ace Glass, Inc. Supplier No. 946311
100mL round bottom flask with a 24/40 center neck, and a 14/20 angled side neck.