Learn More
Ace Glass, Inc. Flask, round bottom, 100mL, 24/40 center, 24/40 side
SDP
Catalog No. 50978936
Encompass_Preferred
100mL round bottom flask with one center and one vertical side neck, both with 24/40 standard taper joints.
Catalog No. 50-978-936 Supplier Ace Glass, Inc. Supplier No. 692807
100mL round bottom flask with one center and one vertical side neck, both with 24/40 standard taper joints.