Learn More
Ace Glass, Inc. Flask, round bottom, 100mL, 24/40 center, #7 Ace-Thred side port
SDP
Catalog No. 50978946
Encompass_Preferred
100mL round bottom flask with 24/40 center and #7 Ace-Thred side port. Supplied with nylon bushing and FETFE o-ring. Bushing allows for use with thermometers or bleed tubes up to 7mm O.D.
Catalog No. 50-978-946 Supplier Ace Glass, Inc. Supplier No. 693630
100mL round bottom flask with 24/40 center and #7 Ace-Thred side port. Supplied with nylon bushing and FETFE o-ring. Bushing allows for use with thermometers or bleed tubes up to 7mm O.D.