Learn More
Ace Glass, Inc. Flask, round bottom, three neck, 12L, 29/42 center, 24/40 sides
SDP
Catalog No. 50973746
Encompass_Preferred
Flask, round bottom, three neck, 12L, 29/42 center, 24/40 sides
Catalog No. 50-973-746 Supplier Ace Glass, Inc. Supplier No. 694869
Flask, round bottom, three neck, 12L, 29/42 center, 24/40 sides