Learn More
Ace Glass, Inc. Flask, round bottom, three neck, 12L, 34/45 center, 29/42 sides
SDP
Catalog No. 50973658
Encompass_Preferred
12L round bottom flask with three vertical necks. Center neck has 34/45 outer joint, and side necks have 29/42 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.
Catalog No. 50-973-658 Supplier Ace Glass, Inc. Supplier No. 694471
12L round bottom flask with three vertical necks. Center neck has 34/45 outer joint, and side necks have 29/42 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.