Learn More
Ace Glass, Inc. Flask, round bottom, three neck, 12L, 55/50 center, 45/50 sides
SDP
Supplier:  Ace Glass, Inc. 694479
Encompass_Preferred
12L round bottom flask with three vertical necks. Center neck has 55/50 outer joint, and side necks have 45/50 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.
Catalog No. 50-973-663