Learn More
Ace Glass, Inc. Flask, round bottom, three neck, 12L, 71/60 center, 45/50 sides
SDP
Catalog No. 50973678
Encompass_Preferred
12L round bottom flask with three vertical necks. Center neck has 71/60 outer joint, and side necks have 45/50 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.
Catalog No. 50-973-678 Supplier Ace Glass, Inc. Supplier No. 6944178
12L round bottom flask with three vertical necks. Center neck has 71/60 outer joint, and side necks have 45/50 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.