Learn More
Ace Glass, Inc. Flask, round, three neck, 15mL, 14/20 center, 14/20 sides
SDP
Catalog No. 50976854
Encompass_Preferred
15mL round bottom flask with three necks for rapid heading. Flask features a 14/20 center neck and two 14/20 angled side necks.
Catalog No. 50-976-854 Supplier Ace Glass, Inc. Supplier No. 946505
15mL round bottom flask with three necks for rapid heading. Flask features a 14/20 center neck and two 14/20 angled side necks.