Learn More
Ace Glass, Inc. Flask, round bottom, three neck, 1L, 24/40 center, 24/40 sides
SDP
Catalog No. 50973637
Encompass_Preferred
1L round bottom flask with three vertical necks. Center neck and side necks have 24/40 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.
Catalog No. 50-973-637 Supplier Ace Glass, Inc. Supplier No. 694424
1L round bottom flask with three vertical necks. Center neck and side necks have 24/40 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.