Learn More
Ace Glass, Inc. Flask, round bottom, 1L, 24/40 center, stopcock septum portside arm
SDP
Catalog No. 50973607
Encompass_Preferred
1L standard wall round bottom flask with a single 24/40 center neck, and a stopcock septum port side arm, for use with air-sensitive material. Flask features a PTFE 2mm bore stopcock. Supplied with a red rubber septum.
Catalog No. 50-973-607 Supplier Ace Glass, Inc. Supplier No. 693429
1L standard wall round bottom flask with a single 24/40 center neck, and a stopcock septum port side arm, for use with air-sensitive material. Flask features a PTFE 2mm bore stopcock. Supplied with a red rubber septum.