Learn More
Ace Glass, Inc. Flask, round bottom, 1L, 29/42 center, 29/42 side
SDP
Catalog No. 50973592
Encompass_Preferred
1L round bottom flask with one center and one vertical side neck, both with 29/42 standard taper joints.
Catalog No. 50-973-592 Supplier Ace Glass, Inc. Supplier No. 692838
1L round bottom flask with one center and one vertical side neck, both with 29/42 standard taper joints.