Learn More
Ace Glass, Inc. Flask, round bottom, four neck, 1L, 34/45 center, (2) 24/40, (1) 10/30 sides
SDP
Catalog No. 50973855
Encompass_Preferred
1L round bottom flask with four necks, arranged three in-line, and one 10/30 joint at 90. Center neck has a 34/45 joint, and vertical side necks have 24/40 joints.
Catalog No. 50-973-855 Supplier Ace Glass, Inc. Supplier No. 695316
1L round bottom flask with four necks, arranged three in-line, and one 10/30 joint at 90. Center neck has a 34/45 joint, and vertical side necks have 24/40 joints.