Learn More
Ace Glass, Inc. Flask, round bottom, 200mL, 24/40 center, beaded tube well
SDP
Catalog No. 50973615
Encompass_Preferred
200mL round bottom, single neck flask with either a beaded tube end port or a beaded tube end well.
Catalog No. 50-973-615 Supplier Ace Glass, Inc. Supplier No. 693566
200mL round bottom, single neck flask with either a beaded tube end port or a beaded tube end well.