Learn More
Ace Glass, Inc. Flask, round bottom, four neck, 22L, 71/60 center, 29/42 sides
SDP
Catalog No. 50973821
Encompass_Preferred
22L round bottom flask with four necks, arranged three in-line and one at 90. Center neck has a 71/60 joint, and angled side necks have 29/42 joints.
Catalog No. 50-973-821 Supplier Ace Glass, Inc. Supplier No. 695289
22L round bottom flask with four necks, arranged three in-line and one at 90. Center neck has a 71/60 joint, and angled side necks have 29/42 joints.