Learn More
Ace Glass, Inc. Flask, round bottom, three neck, 22L, 71/60 center, 45/50 sides
SDP
Catalog No. 50973760
Encompass_Preferred
Flask, round bottom, three neck, 22L, 71/60 center, 45/50 sides
Catalog No. 50-973-760 Supplier Ace Glass, Inc. Supplier No. 694891
Flask, round bottom, three neck, 22L, 71/60 center, 45/50 sides