Learn More
Ace Glass, Inc. Flask, round bottom, three neck, 22L, 71/60 center, 55/50 sides
SDP
Supplier:  Ace Glass, Inc. 694491
Encompass_Preferred
22L round bottom flask with three vertical necks. Center neck has 71/60 outer joint, and side necks have 55/50 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.
Catalog No. 50-973-668