Learn More
Ace Glass, Inc. Flask, round, 250mL, 19/22 center, 14/20 side
SDP
Catalog No. 50976843
Encompass_Preferred
250mL round bottom flask with a 19/22 center neck, and a 14/20 angled side neck.
Catalog No. 50-976-843 Supplier Ace Glass, Inc. Supplier No. 946312
250mL round bottom flask with a 19/22 center neck, and a 14/20 angled side neck.