Learn More
Ace Glass, Inc. Flask, round bottom, four neck, 250mL, 19/22 center, 14/20 sides
SDP
Catalog No. 50976880
Encompass_Preferred
Flask, round bottom, four neck, 250mL, 19/22 center, 14/20 sides
Catalog No. 50-976-880 Supplier Ace Glass, Inc. Supplier No. 946916
Flask, round bottom, four neck, 250mL, 19/22 center, 14/20 sides