Learn More
Ace Glass, Inc. Flask, round, heavy duty, 250mL, 19/22 center, 14/20 side
SDP
Catalog No. 50976847
Encompass_Preferred
250mL heavy wall round bottom flask with a 19/22 center neck, and a 14/20 angled side neck. Heavy wall is approximately 30% heavier than regular wall flasks.
Catalog No. 50-976-847 Supplier Ace Glass, Inc. Supplier No. 9463212
250mL heavy wall round bottom flask with a 19/22 center neck, and a 14/20 angled side neck. Heavy wall is approximately 30% heavier than regular wall flasks.