Learn More
Ace Glass, Inc. Flask, round bottom, four neck, 250mL, 24/40 center, 24/40 sides
SDP
Catalog No. 50973796
Encompass_Preferred
250mL round bottom flask with four necks, arranged three in-line and one at 90. Center neck and angled side necks have 24/40 joints.
Catalog No. 50-973-796 Supplier Ace Glass, Inc. Supplier No. 695201
250mL round bottom flask with four necks, arranged three in-line and one at 90. Center neck and angled side necks have 24/40 joints.