Learn More
Ace Glass, Inc. Flask, round bottom, two neck, 250mL, 24/40 center, 24/40 side
SDP
Catalog No. 50973572
Encompass_Preferred
250mL two neck, round bottom flask with standard joints. Center neck and angled side neck both have 24/40 joints.
Catalog No. 50-973-572 Supplier Ace Glass, Inc. Supplier No. 692708
250mL two neck, round bottom flask with standard joints. Center neck and angled side neck both have 24/40 joints.