Learn More
Ace Glass, Inc. Flask, round bottom, 250mL, 24/40 center, stopcock septum portside arm
SDP
Supplier:  Ace Glass, Inc. 693425
Encompass_Preferred
250mL standard wall round bottom flask with a single 24/40 center neck, and a stopcock septum port side arm, for use with air-sensitive material. Flask features a PTFE 2mm bore stopcock. Supplied with a red rubber septum.
Catalog No. 50-973-605