Learn More
Ace Glass, Inc. Flask, round bottom, three neck, 250mL, 35/25 center, 35/25 sides
SDP
Catalog No. 50973787
Encompass_Preferred
250mL round bottom flask with three vertical necks: (1) 35/25 center neck, (2) 35/25 side necks.
Catalog No. 50-973-787 Supplier Ace Glass, Inc. Supplier No. 695008
250mL round bottom flask with three vertical necks: (1) 35/25 center neck, (2) 35/25 side necks.