Learn More
Ace Glass, Inc. Flask, round, three neck, 25mL, 14/10 cap-seal center and sides
SDP
Catalog No. 5098026
Encompass_Preferred
Flask, round, three neck, 25mL, 14/10 cap-seal center and sides
Catalog No. 50-980-26 Supplier Ace Glass, Inc. Supplier No. 946534
Flask, round, three neck, 25mL, 14/10 cap-seal center and sides