Learn More
Ace Glass, Inc. Flask, round bottom, four neck, 2L, 24/40 center, 24/40 sides
SDP
Catalog No. 50973803
Encompass_Preferred
2L round bottom flask with four necks, arranged three in-line and one at 90. Center neck has a 24/40 joint, and angled side necks have 24/40 joints.
Catalog No. 50-973-803 Supplier Ace Glass, Inc. Supplier No. 695220
2L round bottom flask with four necks, arranged three in-line and one at 90. Center neck has a 24/40 joint, and angled side necks have 24/40 joints.