Learn More
Ace Glass, Inc. Flask, round bottom, three neck, 2L, 34/45 center, 24/40 sides
SDP
Catalog No. 50973644
Encompass_Preferred
2L round bottom flask with three vertical necks. Center neck has 34/45 outer joint, and side necks have 24/40 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades
Catalog No. 50-973-644 Supplier Ace Glass, Inc. Supplier No. 694446
2L round bottom flask with three vertical necks. Center neck has 34/45 outer joint, and side necks have 24/40 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades