Learn More
Ace Glass, Inc. Flask, round bottom, three neck, 2L, 34/45 center, 29/42 sides
SDP
Catalog No. 50973645
Encompass_Preferred
2L round bottom flask with three vertical necks. Center neck has 34/45 outer joint, and side necks have 29/42 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.
Catalog No. 50-973-645 Supplier Ace Glass, Inc. Supplier No. 694447
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
2L round bottom flask with three vertical necks. Center neck has 34/45 outer joint, and side necks have 29/42 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.