Learn More
Ace Glass, Inc. Flask, round bottom, four neck, 3L, 24/40 center, 24/40 sides
SDP
Catalog No. 50973806
Encompass_Preferred
3L round bottom flask with four necks, arranged three in-line and one at 90. Center neck and angled side necks have 24/40 joints.
Catalog No. 50-973-806 Supplier Ace Glass, Inc. Supplier No. 695232
3L round bottom flask with four necks, arranged three in-line and one at 90. Center neck and angled side necks have 24/40 joints.