Learn More
Ace Glass, Inc. Flask, round bottom, four neck, 3L, 24/40 center, (2) 24/40(1) #7 sides
SDP
Catalog No. 50973872
Encompass_Preferred
3L round bottom flask with four necks, arranged three in-line, and one #7 Ace-Thred neck. Center neck and angled side necks have 24/40 joints. Supplied with bushing and FETFE o-ring.
Catalog No. 50-973-872 Supplier Ace Glass, Inc. Supplier No. 695485
3L round bottom flask with four necks, arranged three in-line, and one #7 Ace-Thred neck. Center neck and angled side necks have 24/40 joints. Supplied with bushing and FETFE o-ring.