Learn More
Ace Glass, Inc. Flask, round bottom, three neck, 3L, 29/42 center, 24/40 sides
SDP
Catalog No. 50973647
Encompass_Preferred
3L round bottom flask with three vertical necks. Center neck has 29/42 outer joint, and side necks have 24/40 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.
Catalog No. 50-973-647 Supplier Ace Glass, Inc. Supplier No. 694450
3L round bottom flask with three vertical necks. Center neck has 29/42 outer joint, and side necks have 24/40 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.