Learn More
Ace Glass, Inc. Flask, round bottom, two neck, 3L, 29/42 center, 29/42 side
SDP
Catalog No. 50973576
Encompass_Preferred
3L two neck, round bottom flask with standard joints. Center neck and angled side neck both have 29/42 joints.
Catalog No. 50-973-576 Supplier Ace Glass, Inc. Supplier No. 692758
3L two neck, round bottom flask with standard joints. Center neck and angled side neck both have 29/42 joints.